RPMI 1640 Medium, GlutaMAX™ Supplement
RPMI 1640 Medium, GlutaMAX™ Supplement
RPMI 1640 Medium, GlutaMAX™ Supplement
Gibco™

RPMI 1640 Medium, GlutaMAX™ Supplement

RPMI 1640 Medium (Roswell Park Memorial Institute 1640 Medium) was originally developed to culture human leukemic cells in suspension andRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
6187015010 L
618701435 L
Catalog number 61870150
Price (EUR)
477,00
Each
Quantity:
10 L
Customize this product
Price (EUR)
477,00
Each
RPMI 1640 Medium (Roswell Park Memorial Institute 1640 Medium) was originally developed to culture human leukemic cells in suspension and as a monolayer. RPMI 1640 medium has since been found suitable for a variety of mammalian cells, including HeLa, Jurkat, MCF-7, PC12, PBMC, astrocytes, and carcinomas. We offer a variety of RPMI 1640 modifications for a range of cell culture applications. Find the right formulation using the media selector tool.

This RPMI 1640 is modified as follows:
WithWithout
• GlutaMAX™• HEPES
• Phenol Red

The complete formulation is available.

Using RPMI 1640 Medium
RPMI 1640 medium is unique from other media because it contains the reducing agent glutathione and high concentrations of vitamins. RPMI 1640 medium contains biotin, vitamin B12, and PABA, which are not found in Eagle's Minimal Essential Medium or Dulbecco's Modified Eagle Medium. In addition, the vitamins inositol and choline are present in very high concentrations. RPMI 1640 medium with GlutaMAX™ supplement minimizes toxic ammonia build-up and improves cell viability and growth in an easy-to-use format. RPMI 1640 medium contains no proteins, lipids, or growth factors. Therefore, RPMI 1640 medium requires supplementation, commonly with 10% Fetal Bovine Serum (FBS). RPMI 1640 medium uses a sodium bicarbonate buffer system (2.0 g/L) and therefore requires a 5–10% CO2 environment to maintain physiological pH.

cGMP manufacturing and quality system
RPMI 1640 is manufactured at a cGMP-compliant facility located in Grand Island, New York. The facility is registered with the FDA as a medical device manufacturer and is certified to ISO 13485 standards. For supply chain continuity, we offer an identical RPMI 1640 product made in our Scotland facility (61870-010). This facility is also registered with the FDA as a medical device manufacturer and certified to the ISO 13485 standard.
For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.
Specifications
Cell LineHeLa, Jurkat, MCF-7, PC-12, PBMC, astrocytes, and carcinomas
Cell TypeLeukemic Cells
Concentration1 X
Product LineGibco, GlutaMAX, SynerGy
Product TypeRPMI 1640 Medium (Roswell Park Memorial Institute 1640 Medium)
Quantity10 L
Shelf Life12 Months From Date of Manufacture
Shipping ConditionRoom Temperature
ClassificationAnimal Origin-free
FormLiquid
Serum LevelStandard Serum Supplementation
SterilitySterile-filtered
With AdditivesGlutaMAX, Phenol Red
Without AdditivesNo HEPES, No Sodium Pyruvate
Unit SizeEach
Contents & Storage
Storage conditions: 2-8° C. Protect from light
Shipping conditions: Ambient
Shelf life: 12 months from date of manufacture

Frequently asked questions (FAQs)

How light sensitive is RPMI 1640 media? Should I also be protecting it from LED light?

While we know that different wavelengths of light are worse than others for exposure, we would recommend as a best practice to protect the medium from all forms of light exposure including LEDs, as much as possible to ensure optimal performance, as several components within the medium are light sensitive, such as vitamins.

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.

What is the density (g/L) for RPMI 1640 Medium?

We have specific gravity information for RPMI 1640 Medium: 1.006 kg/L. In this case, the specific gravity is the same as density as the solvent is water.

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.

How long can I keep my media after supplementing with serum?

Generally speaking, media can be used for up to three weeks after supplementation with serum. There are no formal studies to support this, but it is the rule of thumb used by our scientists.

Find additional tips, troubleshooting help, and resources within our Mammalian Cell Culture Basics Support Center.

My medium was shipped at room temperature but it is supposed to be stored refrigerated. Is it okay?

We routinely ship media that require long-term storage in the refrigerator at room temperature. We have done studies on representative media formulations to show that media can be at room temperature for up to a week without a problem.

Find additional tips, troubleshooting help, and resources within our Mammalian Cell Culture Basics Support Center.

Will depletion, absence, or breakdown of essential growth-promoting components such as glutamine or growth factors reduce the growth rate of my culture?

Yes. If you suspect that this is the case, remove the medium and add fresh medium. Alternatively, you can supplement medium with growth-promoting components. It is also possible to substitute GlutaMax I or II for glutamine in the medium to prevent glutamine exhaustion.

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.