This product is only available through your local Thermo Scientific supplier. Learn where to buy >
Orange Filter for myECL™ Imager
Catalog number: 62238
Thermo Scientific™  Related applications: Protein Assays and Analysis | Protein Gel Electrophoresis
Error loading your content!
  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons
Your on-site stock
›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top
Description
The Thermo Scientific 565-620nm bandpass filter is a threaded, 50mm-diameter glass filter. This product is a replacement for the pre-installed orange filter in the myECL Imager, and is used for detection of colorimetric protein stains (silver, coomassie) and wavelengths of common fluorescent nucleic acid gel stains (e.g., ethidium bromide) and protein stains, such as SYPRO* Orange Protein Gel Stain.
More Product Data
Improved Western blot detection using CCD digital imaging
Related Products
myECL™ Imager
For Research Use Only. Not for use in diagnostic procedures.
Specifications
For Use With (Equipment): myECL™ Imager
Product Line: myECL™
Product Size: 1 filter
Type: Filter
Contents & storage
0.0
Documents
Manuals & protocols