DyLight™ 488, Free Acid
Thermo Scientific™

DyLight™ 488, Free Acid

Thermo Scientific DyLight Fluor Free Acids are non-activated forms of selected DyLight Fluorescent Dyes for use as controls and calibrationRead more
Have Questions?
Catalog NumberQuantity
622801 mg
Catalog number 62280
Price (USD)
249.65
Online exclusive
333.00
Save 83.35
Each
Quantity:
1 mg
Request bulk or custom format
Price (USD)
249.65
Online exclusive
333.00
Save 83.35
Each
Thermo Scientific DyLight Fluor Free Acids are non-activated forms of selected DyLight Fluorescent Dyes for use as controls and calibration factors in fluorescence imaging applications. DyLight 488 has excitation and emission peaks at 493 and 518 nm, respectively (±4 nm, in PBS).

These non-activated fluorescent dyes contain the native carboxyl group of the standard dye molecule.

General DyLight Fluorophore Highlights:

• Bright fluorescence—intense emission provides superior sensitivity and requires less conjugate
• Narrow emission spectra—negligible bleed-through between fluorophore channels enables multi-color detection
• Excellent photostability—exceptional resistance to photobleaching enables fluorescence imaging under the most demanding conditions (e.g., structured illumination, 4Pi microscopy)
• Buffer stability—conjugated fluorophores are completely stable at pH 4-9
• Instrument compatibility—excitation and emission spectra correspond with filter sets and laser settings of all popular fluorescence instrumentation

Related Products
DyLight™ 747, Free Acid
DyLight™ 800, Free Acid
For Research Use Only. Not for use in diagnostic procedures.
Specifications
DescriptionDyLight™ 488, Free Acid, 1mg, Solid, DyLight™ 488 Dye, 493/518nm Excitation/Emission Wavelength, <4°C Storage Temperature, Used for Protein Biology, Protein Labeling and Crosslinking, Protein Labeling
Emission518
Excitation493
Label or DyeDyLight™ 488
Product TypeFree Acid
Quantity1 mg
Reactive MoietyCarboxylic Acid
SolubilityDMF (Dimethylformamide)
Label TypeDyLight Fluors
Product LineDyLight
Unit SizeEach
Contents & Storage
Store at < 4°C.