Viper™ TQ Accessories
Thermo Scientific™
Viper™ TQ Accessories
Obtain reproducible chromatographic results using tool-free and easy-to-use Thermo Scientific™ Viper™ TQ UHPLC Fittings.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberFor Use With (Application)Includes
6261.5065AColumn CompatibilityViper TQ Adapter, SST (2 pcs.)
6036.9010Accessory KitReconditioner, union and wrench
6261.5020Reconditioning of Viper TQ fittingsViper TQ reconditioner
6261.5060Connecting Viper TQ capillaries and Viper TQ capillaries with (nano)Viper capillariesViper TQ Union, Titan
6212.0003Fluidic connection for LC systemsViper TQ Tee piece, Titanium
Catalog number 6261.5065A
Price (EUR)
152,00
Pack of 2
For Use With (Application):
Column Compatibility
Includes:
Viper TQ Adapter, SST (2 pcs.)
Price (EUR)
152,00
Pack of 2
Ensure optimal separations and maximum LC performance using virtually zero-dead-volume connections with the Thermo Scientific™ Viper™ TQ UHPLC Fittings. Accessories help complete the installation according to your application needs. Lifespan extending accessories increase the sustainability of the parts using Viper TQ UHPLC Fitting technology.
We offer the following accessories for Viper TQ UHPLC Fittings:
  • Viper TQ Reconditioner to reshape for increased longevity of the seal when connected to multiple columns with non-standard column bore geometry
  • Viper TQ union exclusively designed for connecting Viper TQ UHPLC Fittings and Viper and nanoViper Fingertight Fittings directly without additional dead volume
  • Viper TQ Accessory kit including Viper TQ reconditioner, Viper TQ Union and Viper TQ wrench, for convenient installation of Viper TQ UHPLC Fittings at difficult-to-access places.
  • Adapter for Viper TQ UHPLC Fittings (2 pcs, stainless steel, for columns ≥ 2.1 mm ID) to adapt Viper TQ UHPLC Fittings and Viper Fingertight Fittings to non-standard column bore geometry.
  • Viper TQ Tee-piece designed for connection of Viper TQ capillaries without adding additional dead volume.
Specifications
For Use With (Application)Column Compatibility
IncludesViper TQ Adapter, SST (2 pcs.)
MaterialStainless Steel
Max. Pressure1517 bar
Product LineViper TQ
TypeAdaptor
Unit SizePack of 2