UltiMate™ 3000 Pump Filter Frit
Product Image
Thermo Scientific™
UltiMate™ 3000 Pump Filter Frit
Prevent contaminant particles from reaching your HPLC system by using these filter frits for your Thermo Scientific™ Dionex™ UltiMate™ 3000 Pumps.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberFor Use With (Equipment)Material
6268.0117Ultimate 3000 BM PumpPEEK
6268.0110Dionex UltiMate 3000 SD Pump, Dionex UltiMate 3000 RS PumpStainless Steel
6268.0111Dionex UltiMate 3000 RS Pump, Dionex UltiMate 3000 BM PumpTitanium
Catalog number 6268.0117
Price (MXN)
-
For Use With (Equipment):
Ultimate 3000 BM Pump
Material:
PEEK
Prevent contaminant particles from reaching your HPLC system. Thermo Scientific™ Dionex™ UltiMate™ 3000 Pump Filter Frits remove contaminants that may be present in your solvent supply and prevent them from fouling your LC system. These frits are available in three different materials to match your chromatography needs.
Specifications
For Use With (Equipment)Ultimate 3000 BM Pump
MaterialPEEK
TypeFilter Frit (10μm Porosity)
Unit SizeEach