Vanquish™ HPLC and UHPLC System Accessories
Thermo Scientific™
Vanquish™ HPLC and UHPLC System Accessories
Maximize your liquid chromatography (LC) performance with system accessories best suited for the Thermo Scientific™ Vanquish™ HPLC and UHPLC systems.
Have Questions?
變更視圖buttonViewtableView
產品號碼IncludesFor Use With (Application)Product Line
6268.0130PEEK Mesh HPLC Solvent FiltersEluent filteringVanquish HPLC and UHPLC Systems
6261.1130Viper Guard ConnectorVanquish Binary Pump N or Metering Device, Viper TQ in Viper only ValvesViper, nano Viper, Viper TQ
6036.9003Rear Seal and Needle Wash Tubing Labels for Vanquish SystemsLabeling rear seal and needle wash tubing in Vanquish UHPLC SystemsVanquish HPLC and UHPLC Systems
產品號碼 6268.0130
價格 (TWD)
-
Includes:
PEEK Mesh HPLC Solvent Filters
For Use With (Application):
Eluent filtering
Product Line:
Vanquish HPLC and UHPLC Systems
Extend the compatibility and lifespan of nanoViper™ Fignertight Fitting Systems and Viper™ TQ UHPLC Fittings with the Viper Guard Connector. Add the Thermo Scientific™ PEEK Mesh HPLC solvent filter and Rear Seal and Needle Wash Tubing Labels for Vanquish LC Systems for enhance user-friendliness.
Optimizing the chromatographic performance and user-friendliness with accessories suited best for Vanquish HPLC and UHPLC systems.
規格
For Use With (Application)Eluent filtering
IncludesPEEK Mesh HPLC Solvent Filters
MaterialPEEK
For Use With (Equipment)Vanquish Horizon, Flex UHPLC and Core HPLC Systems
Product LineVanquish HPLC and UHPLC Systems
TypeSolvent Filters
Unit SizeEach