Vanquish™ Pump Mixers
Actual product may vary
Vanquish™ Pump Mixers
Use these biocompatible pump mixers in Vanquish UHPLC systems for optimal mixing, including difficult agents such as TFA.
Have Questions?
보기 방식 변경buttonViewtableView
카탈로그 번호설명부피(미터법)
6268.5310Mixer Set, Vanquish H Pump400μL
6268.5120Mixer Set, Vanquish H Pump200 μL
6044.5110Static Mixer, Vanquish H/F Pumps150 μL
6044.5310Static Mixer, Vanquish H/F Pumps350 μL
6044.5018Mixer Set, Vanquish H Pump35 μL
6044.3870Mixer Set, Vanquish F Pumps35 μL
6044.5100Mixer Set, Vanquish F Pumps100 μL
6044.5750AStatic Mixer, Vanquish F Pumps750 μL
6044.5450AStatic Mixer, Vanquish F Pumps1500 μL
6044.5026Capillary Mixer, Vanquish F Pumps50 μL
카탈로그 번호 6268.5310
제품 가격(KRW)
-
설명:
Mixer Set, Vanquish H Pump
부피(미터법):
400μL
Use the biocompatible Thermo Scientific™ Vanquish™ Pump Mixers with Vanquish binary, quaternary, or dual gradient UHPLC pumps to ensure the highest possible mixing efficiency for ultrahigh resolution and ultrafast separations. These mixers accomplish complete mixing of the HPLC gradient elution to achieve a smooth baseline with high signal-to-noise ratios, and accurate peak identification. Vanquish Pump Mixers are designed to reliably provide the highest possible performance over a wide application range with varying solvent ratios, solvent miscibilities, flow rates.
사양
설명Mixer Set, Vanquish H Pump
포함Static mixer: 350 μL<br /> Capillary mixer: 50 μL
부피(미터법)400μL
용도 (장비)Vanquish Binary Pump H
제품라인Vanquish
유형Mixer Set
Unit SizeEach