See more signal from immunofluorescence images with Prolong® Diamond. Learn more.

Goat Anti-Human IgA - BT

Catalog number: 627440

Invitrogen™  Related applications: Secondary Detection

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Immunogen: Human IgA

Specificity: Human IgA

Applications: ELISA, Immunohistochemistry (FFPE), Immunofluorescence, Immunoprecipitation, WB
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Reactivity: Human
Target Isotype: IgA
Host Species: Goat
Antibody Fragment: Whole Antibody
Label or Dye: Biotin
Product Size: 1 mL
Shipping Condition: Wet Ice

Contents & storage

Store in refrigerator (2–8°C).

Documents

Manuals & protocols

Recommended products