Product Image

PrioCHECK™ MAP Ab 2.0 Plate Kit

Have Questions?
Catalog number 63328
Price (MXN)
-
Note: The name of this product has changed. It was previously called PARACHEK 2 30 PLATES.

PARACHEK® 2 is a solid phase, indirect enzyme immunoassay (EIA) to detect antibodies to M. paratuberculosis in animal sera and milk of cattle, sheep, and goats. The PARACHEK® 2 procedure for 96 determinations (including controls) takes approximately 80 min and consists of four steps. In the first step, the samples are diluted and incubated in a diluent buffer containing M. phlei to remove cross-reacting antibodies. In the second step, specific antibodies present in the diluted samples bind to M. paratuberculosis antigens coated to the wells of a microtiterplate. Unreacted proteins are removed by washing. A secondary antibody that is conjugated to an enzyme that generates a color signal is added in the third step. In the fourth step, the presence of specific antibodies is visualized by adding the enzyme substrate. The intensity of the color signal is proportional to the amount of antibodies in the sample.
Specifications
Assay SensitivitySee product documentation
Gene IDNon-Gene
Gene SymbolNon-Gene
Immunoassay Kit FormatVarious Other
Product LinePARACHEK®,Prionics™
Product Size2760 samples
Protein FamilyOther Proteins
Target (Kit Named)PARACHEK™ 2 Kit
Target Organism ClassMycobacterium avium spp paratuberculosis (MAP)
Target Species (Named)Bovine
Target Species (Validated)Bovine,Goat,Sheep
Unit Size2760 samples
Contents & Storage
30 plates