Nalgene™ Polycarbonate Safety Splash Shield
Nalgene™ Polycarbonate Safety Splash Shield
Nalgene™ Polycarbonate Safety Splash Shield
Nalgene™ Polycarbonate Safety Splash Shield
Nalgene™ Polycarbonate Safety Splash Shield
Thermo Scientific™

Nalgene™ Polycarbonate Safety Splash Shield

Simultaneously maintain safety and a clear field of vision by using this safety splash shield. Provides protection from splashes, aerosols and flying debris.
Have Questions?
Catalog NumberDescriptionLength (English)Width (English)
6356-0001PK
also known as 6356-0001
Safety Splash Shield with acetal flexible arm; PC 21.6 x 27.9 cm12 in.8.5 in.
Catalog number 6356-0001PK
also known as 6356-0001
Price (TWD)
-
Description:
Safety Splash Shield with acetal flexible arm; PC 21.6 x 27.9 cm
Length (English):
12 in.
Width (English):
8.5 in.

Simultaneously maintain safety and a clear field of vision by using the Thermo Scientific™ Nalgene™ Polycarbonate Safety Splash Shield. This shield attaches easily to benchtop shelving, machinery or other magnetic surfaces for convenient protection from splashes, aerosols and flying debris. This durable, scratch-resistant 1/8 in. (3mm) thick polycarbonate shield protects the user and features an easy to maneuver arm. Meets OSHA 29CFR1910.1030 standard.

  • Flexible 14 inch arm makes it easy to maneuver shield over samples
  • Magnetic base firmly grips to magnetic surfaces letting you position the shield horizontally (flat), vertically or on an overhead surface
  • Provides UV protection from 200-360nm for added versatility
  • Biohazard/Transparent
Warning!: Do not use with radioisotopes. Use Nalgene beta-radiation shields when working with beta-radioisotopes. Do not autoclave.

Please recycle the corrugate cartons and any LDPE bags your Nalgene products are delivered in. 

 

 

Specifications
Certifications/ComplianceMeets OSHA 29CFR1910.1030 standard
DescriptionSafety Splash Shield with acetal flexible arm; PC 21.6 x 27.9 cm
Length (English)12 in.
No. per Case4/Cs.
No. per Pack1 Ea.
Thickness (English)0.12 in.
Thickness (Metric)3 mm
Width (English)8.5 in.
Width (Metric)21.5 cm
Lens MaterialPolycarbonate
Lens TintClear
MaterialPolycarbonate (Lens)
Product TypeSafety Splash Shield
UV ProtectionYes
Unit SizeEach