para-JEM™ Bottle Carrying Rack
para-JEM™ Bottle Carrying Rack
para-JEM™ Bottle Carrying Rack
Thermo Scientific™
para-JEM™ Bottle Carrying Rack
Safely and conveniently transport Thermo Scientific™ para-JEM™ reagents for automated detection of M. avium ssp. paratuberculosis.
Have Questions?
Catalog NumberPackaging
6423-3024 Bottles/Pk.
Catalog number 6423-30
Price (MXN)
-
Packaging:
24 Bottles/Pk.
Safely and conveniently transport Thermo Scientific™ para-JEM™ reagents for automated detection of M. avium ssp. paratuberculosis with the para-JEM Bottle Carrying Rack.
Solid medium culture used to be the standard procedure for detecting M. avium ssp. paratuberculosis (MAP), the causative agent of Johne's Disease, in ruminant fecal samples before para-JEM reagents. Due to the bacterium's long generation time, it can take up to 12 weeks of incubation on solid medium before the bacteria are detected. The para-JEM System is a unique, liquid culture method that combines a full line of reagents with the powerful Thermo Scientific™ VersaTREK™ Automated Microbial Detection System for faster, automated detection of MAP.
The para-JEM System includes a number of unique features:
  • Detects faster—able to recover MAP in as little as 2 to 3 weeks, much earlier than conventional methods*
  • Provides increased sensitivity over conventional methods*
  • Provides semi-quantitation for low, medium and high shedders based on time-to-detection*
  • Reduces labor with fewer false positives, false negatives and contaminants*
  • Improves workflow with most tests concluded at six weeks for high, medium and many low shedders*
Utilize para-JEM reagents with the fully automated VersaTREK Microbial Detection System, which automatically incubates and continuously monitors culture bottles for accurate, reliable organism detection, as well as enhanced ergonomics and workflow versatility.
Not all products are available for sale in all territories. Please inquire.
VersaTREK, Sensititre™, Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products with a comprehensive line of media and diagnostic products, providing a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
*Cited as a conclusion from an abstract presented at the 43rd Annual AAVLD Meeting - A New Liquid Culture Method, the TREK ESP Culture System II, for the Rapid Detection of Mycobacterium avium subsp. paratuberculosis in Bovine Fecal Samples, S.J.Shin, et. al, College of Veterinary Medicine Diagnostic Laboratory, Cornell University
Specifications
Descriptionpara-JEM Bottle Carrying Rack
Packaging24 Bottles/Pk.
Product TypeRack
Unit SizeEach