Product Image
Invitrogen™

eBioscience™ Fura-2 AM Dye

Fura-2 is a calcium imaging dye that binds to free Ca2+. Fura-2 AM is the cell-permeable acetoxymethyl (AM) ester formRead more
Have Questions?
Catalog NumberQuantity
65-0858-391 mg
Catalog number 65-0858-39
Price (KRW)
338,000
Online offer
Ends: 31-Dec-2026
397,000
Save 59,000 (15%)
Each
Quantity:
1 mg
Price (KRW)
338,000
Online offer
Ends: 31-Dec-2026
397,000
Save 59,000 (15%)
Each
Fura-2 is a calcium imaging dye that binds to free Ca2+. Fura-2 AM is the cell-permeable acetoxymethyl (AM) ester form of Fura-2. Fura-2 is the preferred dye for ratiometric imaging microscopy with digital image analysis, especially when the alteration of excitation wavelengths is more practical than the detection of multiple emission wavelengths. Upon binding Ca2+, the excitation spectrum of Fura-2 shifts to shorter wavelengths between 300 and 400 nm, while the peak emission remains steady around 510 nm. The Kd of Fura-2 is highly dependent on pH, temperature, ionic strength and viscosity of the cytosol, thus great care should be taken when non-standard conditions are used.

Molecular weight: 1001.86
Peak excitation: variable depending on the concentration of free Ca2+, between 300 and 400 nm
Peak emission: 510 nm

Fura-2 AM should be reconstituted in high-quality, freshly opened DMSO. Once reconstituted, it should be protected from light and stored at -20°C. Avoid freeze-thawing.

Reported Application
Microscopy, Cell Labeling

For Research Use Only. Not for use in diagnostic procedures.
Specifications
Detection MethodFluorescence
Emission Wavelength300 to 400/510
Quantity1 mg
For Use With (Equipment)Fluorescence Microscope, Flow Cytometer
Product LineeBioscience
Product TypeDye
Unit SizeEach