Product Image
Invitrogen™

eBioscience™ Fixable Viability Dye eFluor™ 506

Fixable Viability Dye eFluor™ 506 is a viability dye that can be used to irreversibly label dead cells prior toLeia mais
Have Questions?
Alterar visualizaçãobuttonViewtableView
Número do catálogoQuantity
65-0866-18500 Tests
65-0866-14100 Tests
Número do catálogo 65-0866-18
Preço (BRL)
3.772,35
Each
Quantity:
500 Tests
Preço (BRL)
3.772,35
Each
Fixable Viability Dye eFluor™ 506 is a viability dye that can be used to irreversibly label dead cells prior to cryopreservation, fixation and/or permeabilization procedures. Unlike 7-AAD and propidium iodide, cells labeled with Fixable Viability Dyes can be washed, fixed, permabilized, and stained for intracellular antigens without any loss of staining intensity of the dead cells. Thus, using Fixable Viability Dyes allows dead cells to be excluded from analysis when intracellular targets are being studied. Fixable Viability Dyes may be used to label cells from all species.

Fixable Viability Dye eFluor™ 506 can be excited by the violet (405 nm) laser line and has a peak emission of 506 nm that can be detected using a 510/50 band pass filter (equivalent to AmCyan). Please make sure that your instrument is capable of detecting this dye. For compensation, it is recommended to use a sample of the cells of interest stained with the Fixable Viability Dye. If the percentage of dead cells is expected to be less than 5%, then it is recommended to take a small aliquot of cells and heat them at 65°C for 1 minute then immediately place on ice for 1 minute. After this treatment, the heat-killed cells can be combined 1:1 with live cells and then stained with the Fixable Viability Dye.

Fixable Viability Dye eFluor™ 506 is supplied as a pre-diluted solution prepared in high-quality, anhydrous DMSO. It should be protected from light and moisture. Store at less than or equal to -70°C with dessicant. It may be freeze-thawed up to 20 times. Allow vial to equilibrate to room temperature before opening.

Reported Application
Flow Cytometric Analysis

For Research Use Only. Not for use in diagnostic procedures.
Especificações
Detection MethodFluorescence
Emission Wavelength405/506
Quantity500 Tests
ColorCyan
For Use With (Application)Viability Assay
For Use With (Equipment)Flow Cytometer
Product LineeBioscience
Product TypeDye
Unit SizeEach

Frequently asked questions (FAQs)

Can Fixable Viability Dyes be used with the eBioscience BrdU Staining kits for Flow Cytometry?

Yes, a Fixable Viability Dye can be added to samples after BrdU labeling and before fixation with the BrdU Staining Buffer. See the protocol for additional information.

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.