Product Image
Have Questions?
Catalog number 650-08-9630XL
Price (CLP)
-
Frequently asked questions (FAQs)
What accessories are available for the Versette Automated Liquid Handler?
Here are the accessories available for the Versette Automated Liquid Handler:
- Three pipetting heads (volume range 0.5 µL - 300 µL)
- Peristaltic pump including non-contact reservoir fill
- Tip wash stations for 96/384-well formats
- Four serial dilution magazines for 96/384-well applications