Dynabeads™ Streptavidin for In Vitro Transcription
Invitrogen™

Dynabeads™ Streptavidin for In Vitro Transcription

Dynabeads Streptavidin for In Vitro Transcription offers an innovative technology for simple, flexible, and automation-ready mRNA synthesis in scales ranging from microgram to gram.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberVolume (Metric)
65005D2 mL
65006D10 mL
Catalog number 65005D
Price (USD)
718.00
Each
Volume (Metric):
2 mL
Price (USD)
718.00
Each

Dynabeads Streptavidin for In Vitro Transcription offer an innovative technology for simple, flexible, and automatable mRNA synthesis, in scales ranging from micrograms to grams. They are particularly suitable where flexibility and high quality are required, such as in the discovery phase of RNA-based vaccines and therapeutics development.

The 1-μm monosized, monodispersed, paramagnetic beads are covalently coupled with streptavidin molecules, for binding to biotinylated DNA template. The workflow is designed to minimize the effort and cost of plasmid preparation, while enabling reuse of template-bead complex in successive solid-phase in vitro transcription (IVT) reactions (figure below).

Features of Dynabeads Streptavidin for In Vitro Transcription include:

  • Flexibility – choose DNA template for IVT with flexibility of design and amplification
  • Efficiency – reuse DNA template and beads at least six times to help reduce cost and eliminate/minimize plasmid prep hands-on time and risk of bacterial host DNA transfer
  • Scalability – swap between manual and automated workflows to match required scale (micrograms to grams of mRNA) and throughput
  • Quality – help ensure a safe and flexible supply of magnetic beads to navigate from RNA-based drug discovery research to commercial manufacturing with counterpart solutions (available via the OEM supply) without incurring extra time or costs from switching technology or raw material

Overview of the workflow
A biotinylated template is made by PCR amplification of the target sequence in a plasmid construct or in a synthetic DNA construct using a biotinylated forward primer and a non-biotinylated reverse primer. As an alternative to PCR amplification, a linearized plasmid can be biotinylated by a fill-in reaction of biotin-dUTP in the 5' overhang sequence, provided the correct plasmid design. The biotinylated template is then immobilized directly to the Dynabeads Streptavidin for In Vitro Transcription beads without the need for prior purification. The bead-bound template is used directly in the IVT, then removed from the synthesized mRNA by magnetic separation (Figure 1). The template can be reused (at least six times) in consecutive IVT reactions (figure below).

Solid-phase IVT is compatible with commonly used reagents such as MEGAscript T7 Transcription Kit, mMESSAGE mMACHINE T7 Transcription Kit, and mMESSAGE mMACHINE T7 ULTRA Transcription Kit, based on your needs and strategies for capping and polyadenylation.

The beads are offered here as a stand-alone product. They are also included in the Dynabeads In Vitro Transcription and RNA Purification Kit.

Automation-ready for use with KingFisher purification systems
The 1-μm magnetic beads are ideal for high-throughput screening. The process can be automated using liquid handlers or any of our KingFisher sample purification systems. The scripts for KingFisher Flex and KingFisher Apex automated purification workflows are available from our website.

Commercial supply
To learn more about counterpart products and support documentation available for GMP-compliant manufacturing of mRNA vaccines and therapeutics, visit: thermofisher.com/dynabeadsmrna

For Research Use Only. Not for use in diagnostic procedures.
Specifications
Certifications/ComplianceISO9001 and ISO13485
ColorBrown
Concentration10 mg/mL
Diameter (Metric)1 μm
For Use With (Application)RNA Purification
For Use With (Equipment)KingFisher™ Sample Purification System, DynaMag™ magnets
High-throughput CompatibilityHigh-throughput Compatible
Ligand TypeBiotin
MaterialMagnetic Polystyrene
Product LineDynabeads
Product TypeStreptavidin Magnetic Beads
Quantity2 mL
Shelf Life48 Months (from date of manufacture)
Shipping ConditionAmbient Temperature
UniformityMonosized 1 μm (CV <5%)
Volume (Metric)2 mL
Isolation TechnologyMagnetic Bead
Unit SizeEach
Contents & Storage
Dynabeads Streptavidin for In Vitro Transcription is supplied in PBS, pH 7.4, with 20% ethanol and 0.01% Tween-20. Store at 2°C to 8°C.

Frequently asked questions (FAQs)

What is the concentration of Dynabeads Streptavidin for In Vitro Transcription (Cat. Nos. 65005D, 65006D)?

The concentration of Dynabeads Streptavidin for In Vitro Transcription is 10 mg/mL, approximately 7-9 x 109 beads/mL.
Note: The numbers will be within this range, but vary from batch to batch.

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.