Thermo Scientific™
Richard-Allan Scientific™ Neg-50™ Frozen Section Medium
 Related applications:
Anatomical Pathology
Yield high-quality sections down to -50°C without chatter using Thermo Scientific™ Richard-Allan Scientific™ Neg-50™ Frozen Section Medium, a water-soluble frozen section medium.
Error loading your content!
  Catalog number Price ({{currency}}) Availability Qty
{{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}} Add To Cart
{{addItemToList.successMessage}} {{addItemToList.listName}}
Back to top
Description
  • Leaves no residue after staining
  • Compatible with the Microm Automatic Cryo Approach (ACA) system
Ordering Information:
Available in five colors for easy identification of specimens: pink, blue, green, yellow, and orange. Multi-packs contain one of each color.
Documents
Manuals & protocols