Thermo Scientific™
Richard-Allan Scientific™ Neg-50™ Frozen Section Medium
Related applications:
Anatomical Pathology
Yield high-quality sections down to -50°C without chatter using Thermo Scientific™ Richard-Allan Scientific™ Neg-50™ Frozen Section Medium, a water-soluble frozen section medium.
Back to top
  • Leaves no residue after staining
  • Compatible with the Microm Automatic Cryo Approach (ACA) system
Ordering Information:
Available in five colors for easy identification of specimens: pink, blue, green, yellow, and orange. Multi-packs contain one of each color.