Thermo Scientific™
eVol™ XR Dispensing System
 Related applications:
Industrial Chromatography
Conventional pipetting can sometimes lead to errors and rework, depending on which technician is doing the work. Now there's a way to eliminate dispensing variability and improve efficiency. The Thermo Scientific™ eVol™ Dispensing System is a unique, hand-held digital dispenser that accurately and reproducibly performs a wide variety of liquid handling procedures. The easy-to-use device couples an analytical syringe to a programmable electronic controller, enabling highly reproducible dispensing for all users.
Error loading your content!
  Catalog number Price ({{currency}}) Availability Qty
{{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}} Add To Cart
{{addItemToList.successMessage}} {{addItemToList.listName}}
Back to top
Description
The eVol Dispensing System is ideal for laboratories that need to repeatedly dispense nonaqueous liquids, offering greater accuracy and reproducibility for applications such as:
  • Routine dispensing of volatile solvents, hazardous, corrosive or viscous chemicals
  • Preparation of calibration standards
  • Addition of internal standards
The system unifies two precision devices: a digitally controlled electronic drive, and a Thermo Scientific™ XCHANGE™ enabled analytical syringe.
Repeatable Performance for Higher Confidence in Your Results
  • Can be calibrated easily by a user to ensure validity of results
  • Intuitive, easy-to-use interface with touch wheel and full-color screen
  • Complies with GLP and GMP guidelines
Exceptional Versatility and Functionality
  • Integrated XCHANGE coupling allows syringes to be quickly and easily changed, preventing possible cross-contamination
  • Addition of Thermo Scientific™ MEPS™ (Micro Extraction by Packed Sorbent) technology to an XCHANGE syringe enables automated sample preparation by SPE in a hand-held system
Documents
Manuals & protocols