Thermo Scientific™
eVol™ XR Dispensing System Syringes
Catalog number: 66002-023
Rely on exceptional versatility and functionality when using Thermo Scientific™ eVol™ XCHANGE Syringes. For use with the unique, hand-held eVol XR Dispensing System, the syringes are available in sizes from 5 to 1000μL. Integrated XCHANGE™ coupling allows you to change syringes quickly and easily. The addition of MEPS to XCHANGE syringe offers automated sample preparation by SPE in a handheld system. The eVol XR Dispensing System accurately and reproducibly performs a wide variety of liquid handling procedures