Thermo Scientific™
eVol™ XR Sample Dispensing System Kit
Catalog number: 66002-024
Thermo Scientific™
eVol™ XR Sample Dispensing System Kit
Catalog number: 66002-024
Get up and running quickly. The Thermo Scientific™ eVol™ Sample Dispensing System Kit includes an eVol XR dispensing system and three syringes (5, 50, and 500μL). The eVol Dispensing System is a unique, hand-held digital dispenser that accurately and reproducibly performs a wide variety of liquid handling procedures. The easy-to-use device couples an analytical syringe to a programmable electronic controller, enabling highly reproducible dispensing for all users.
Catalog Number
66002-024
Unit Size
Each
Description
Sample Dispensing System Kit
For Use With (Application)
Routine Dispensing of Volatile Solvents, Hazardous, Corrosive or Viscous Chemicals
Price (USD)
Price: 2,101.00
Your Price:
Full specifications
Quantity
DescriptionSample Dispensing System Kit
For Use With (Application)Routine Dispensing of Volatile Solvents, Hazardous, Corrosive or Viscous Chemicals
Product LineHyperSep
TypeDispensingSystem
For Use With (Equipment)Routine Dispensing of Volatile Solvents, Hazardous, Corrosiveor Viscous Chemicals
Unit SizeEach
Showing 1 of 1
Catalog NumberSpecificationsUnit SizeDescriptionFor Use With (Application)Price (USD)Quantity
66002-024Full specifications
EachSample Dispensing System KitRoutine Dispensing of Volatile Solvents, Hazardous, Corrosive or Viscous Chemicals
Price: 2,101.00
Your Price:
DescriptionSample Dispensing System Kit
For Use With (Application)Routine Dispensing of Volatile Solvents, Hazardous, Corrosive or Viscous Chemicals
Product LineHyperSep
TypeDispensingSystem
For Use With (Equipment)Routine Dispensing of Volatile Solvents, Hazardous, Corrosiveor Viscous Chemicals
Unit SizeEach
Showing 1 of 1
The eVol Dispensing System is ideal for laboratories that need to repeatedly dispense nonaqueous liquids, offering greater accuracy and reproducibility for applications such as:
  • Routine dispensing of volatile solvents, hazardous, corrosive or viscous chemicals
  • Preparation of calibration standards
  • Addition of internal standards
The system unifies two precision devices: a digitally controlled electronic drive, and a Thermo Scientific™ XCHANGE™ enabled analytical syringe.
Repeatable Performance for Higher Confidence in Your Results
  • Can be calibrated easily by a user to ensure validity of results
  • Intuitive, easy-to-use interface with touch wheel and full-color screen
  • Complies with GLP and GMP guidelines
Exceptional Versatility and Functionality
  • Integrated XCHANGE coupling allows syringes to be quickly and easily changed, preventing possible cross-contamination
  • Addition of Thermo Scientific™ MEPS™ (Micro Extraction by Packed Sorbent) technology to an XCHANGE syringe enables automated sample preparation by SPE in a hand-held system
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Figures