Thermo Scientific™
eVol™ XR Dispensing System Plungers
Catalog number: 66002-041
Thermo Scientific™
eVol™ XR Dispensing System Plungers
Catalog number: 66002-041
Use these Thermo Scientific™ eVol™ Dispensing System Plungers with the respective size syringe on the eVol Dispensing System. Like the syringes, these plungers are available in 5 to 1000μL sizes. The unique, hand-held eVol Dispensing System accurately and reproducibly performs a wide variety of liquid handling procedures. The easy-to-use device couples an analytical syringe to a programmable electronic controller, enabling highly reproducible dispensing for all users.
Catalog Number
66002-041
Unit Size
Each
Description
Replacment plunger for 1000µL eVol system, 1 pk
Price (USD)
Full specifications
DescriptionReplacment plunger for 1000µL eVol system, 1 pk
Product LineHyperSep
TypeDispensing System Plunger
For Use With (Equipment)Electronic Dispensing System
Unit SizeEach
Catalog Number
Unit Size
Each
Description
Replacment plunger for 5µL eVol system, 1 pk
Price (USD)
Full specifications
DescriptionReplacment plunger for 5µL eVol system, 1 pk
Product LineHyperSep
TypeDispensing System Plunger
For Use With (Equipment)Electronic Dispensing System
Unit SizeEach
Catalog Number
Unit Size
Each
Description
Replacment plunger for 500µL eVol system, 1 pk
Price (USD)
Full specifications
DescriptionReplacment plunger for 500µL eVol system, 1 pk
Product LineHyperSep
TypeDispensing System Plunger
For Use With (Equipment)Electronic Dispensing System
Unit SizeEach
Showing 3 of 3
Catalog NumberSpecificationsUnit SizeDescriptionPrice (USD)
66002-041Full specifications
EachReplacment plunger for 1000µL eVol system, 1 pkRequest A Quote
DescriptionReplacment plunger for 1000µL eVol system, 1 pk
Product LineHyperSep
TypeDispensing System Plunger
For Use With (Equipment)Electronic Dispensing System
Unit SizeEach
66002-029Full specifications
EachReplacment plunger for 5µL eVol system, 1 pkRequest A Quote
DescriptionReplacment plunger for 5µL eVol system, 1 pk
Product LineHyperSep
TypeDispensing System Plunger
For Use With (Equipment)Electronic Dispensing System
Unit SizeEach
66002-031Full specifications
EachReplacment plunger for 500µL eVol system, 1 pkRequest A Quote
DescriptionReplacment plunger for 500µL eVol system, 1 pk
Product LineHyperSep
TypeDispensing System Plunger
For Use With (Equipment)Electronic Dispensing System
Unit SizeEach
Showing 3 of 3
The eVol Dispensing System is ideal for laboratories that need to repeatedly dispense nonaqueous liquids, offering greater accuracy and reproducibility for applications such as:
  • Routine dispensing of volatile solvents, hazardous, corrosive or viscous chemicals
  • Preparation of calibration standards
  • Addition of internal standards
The system unifies two precision devices: a digitally controlled electronic drive, and a Thermo Scientific™ XCHANGE™ enabled analytical syringe.
Repeatable Performance for Higher Confidence in Your Results
  • Can be calibrated easily by a user to ensure validity of results
  • Intuitive, easy-to-use interface with touch wheel and full-color screen
  • Complies with GLP and GMP guidelines
Exceptional Versatility and Functionality
  • Integrated XCHANGE coupling allows syringes to be quickly and easily changed, preventing possible cross-contamination
  • Addition of Thermo Scientific™ MEPS™ (Micro Extraction by Packed Sorbent) technology to an XCHANGE syringe enables automated sample preparation by SPE in a hand-held system
Figures