Thermo Scientific™
Richard-Allan Scientific™ Xylene
 Related applications: Anatomical Pathology
Obtain superior lipid extraction, optimal paraffin infiltration, and excellent deparaffinization and clearing during staining with Thermo Scientific™ Richard-Allan Scientific™ Xylene.
Error loading your content!
  Catalog number Price ({{currency}}) Availability Qty
{{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}} Add To Cart
{{addItemToList.successMessage}} {{addItemToList.listName}}
Back to top
Description
  • For use in all tissue processors and automatic stainers
  • Contains the lowest level of benzene, a known carcinogen
  • Strict quality assurance, including gas chromatography, is performed on each lot to guarantee a consistent product
Documents
Manuals & protocols