Product Image
Thermo Scientific™
Laboratory Refrigerator and Freezer Casters
Achieve higher elevation on high-performance refrigerators and freezers with these laboratory refrigerator and freezer casters. Casters offer a 2 or 3 in. height increase.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberFor Use With (Equipment)
6694-145.8 cu. ft. Units
60414.9, 11.5, 23.3, 29.2, 51.1, 78.8 cu. ft. Units
6041-14.9, 11.5, 23.3, 29.2, 51.1, 78.8 cu. ft. Units
609445.8 cu. ft. Units
669445.8 cu. ft. Refrigerators with Sliding Glass Doors
69834.9, 11.5, 23.3, 29.2, 45.8, 51.1, 78.8 cu. ft. Freezers
Catalog number 6694-1
Price (TWD)
-
For Use With (Equipment):
45.8 cu. ft. Units
Achieve higher elevation on high-performance refrigerators and freezers with Thermo Scientific™ Laboratory Refrigerator and Freezer Casters. Casters offer a 2 or 3 in. height increase for Thermo Scientific™ Revco™ , Forma™ and Puffer Hubbard™ laboratory refrigerators and freezers.
  • An option for replacement on most models
  • Factory-installed when ordered with unit; customer-installed if ordered separately
  • 2 in. (50mm) casters are standard on high-performance refrigerators and freezers
  • Optional 3 in. (76mm) casters are shipped with the cabinet but must be installed upon delivery
Ordering Information:
  • 6041: 2 in. casters (50mm), set of four
  • 6041-1: 3 in. casters (76mm), set of four
  • 6983: Locking casters for -30°C freezers only, includes two nonlocking 2 in. (50mm) casters
Specifications
For Use With (Equipment)45.8 cu. ft. Units
Type3 in. Dual Wheel
Capacity (English)45.8 cu. ft.
Capacity (Metric)1296.9 L
Temperature Range-30°C
Unit SizeEach