Freezer and Refrigerator Shelves
Actual product may vary
Freezer and Refrigerator Shelves
Freezer and Refrigerator Shelves
Freezer and Refrigerator Shelves
Thermo Scientific™
Freezer and Refrigerator Shelves
Maximize lab storage with Thermo Scientific™ Freezer and Refrigerator Shelves, designed for use as replacement shelves in high-performance laboratory refrigerators, freezers and ULTs.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberFor Use With (Equipment)Type
6696-123.3, 45.8, 51.1 cu.ft. UnitsFull Solid Shelves
669529.2 cu. ft. UnitsOpen Wire Shelves
6695-129.2 cu. ft. High-performance UnitsOpen Wire Shelves
669623.3, 45.8, 51.1, 78.8 cu. ft. UnitsOpen Wire Shelves
669729.2 cu. ft. UnitsOpen Wire Shelves
669851.1 cu. ft. UnitsOpen Wire Shelves
Catalog number 6696-1
Price (KRW)
-
For Use With (Equipment):
23.3, 45.8, 51.1 cu.ft. Units
Type:
Full Solid Shelves
Maximize lab storage with Thermo Scientific™ Freezer and Refrigerator Shelves, designed for use as replacement shelves in high-performance laboratory refrigerators, freezers and ULTs.
  • Epoxy-coated open-wire shelves
  • Full- and half-depth sizes
  • Undercounter units use full-depth shelves only
  • Warranty varies by country, please contact us for details
Specifications
Exterior FinishStainless Steel
For Use With (Equipment)23.3, 45.8, 51.1 cu.ft. Units
HoldsGeneral Freezer Items
TypeFull Solid Shelves
WarrantyWarranty varies by country, please contact us for details
Unit SizeEach