Gibco™

Resurge™ CD1 Supplement, without glucose and L-glutamine

Catalog number:  670012

 Related applications:

Mammalian Cell Culture

| Viral Vaccine Production

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Resurge CD1 Supplement, without glucose and L-glutamine, is a chemically defined supplement formulation to complement a variety of base media. This supplement was developed using several host cell lines and in various culture systems, including shaker flasks and bench-scale bioreactors. Resurge CD1 Supplement, without glucose and L-glutamine, was developed for use in CHO mammalian culture systems and can be used either as a media supplement or a culture system feed.

Specifications

Cell Line: CHO
Classification: Animal Origin-Free, Chemically Defined, Protein-Free
Form: Powder
Format: Bottle(s)
Product Line: Resurge™
Product Size: 1 kg

Contents & storage

Storage conditions: 2°C to 8°C. Store in cool, dry conditions in well-sealed containers.

Documents

Manuals & protocols