Gibco™

Resurge™ CD2 Supplement, without glucose and L-glutamine

Catalog number:  670016

Related applications:

Cell Culture

| Bioprocessing

Back to top

Description

Resurge CD2 Supplement, without glucose and L-glutamine, is a chemically defined supplement formulation to complement a variety of base media. This supplement was developed using several host cell lines and in various culture systems, including shaker flasks and bench-scale bioreactors. Resurge CD2 Supplement, without glucose and L-glutamine, was developed for use in CHO mammalian culture systems and can be used either as a media supplement or a culture system feed.

Specifications

Cell Line: CHO
Classification: Animal Origin-Free, Chemically Defined, Protein-Free
Format: Bottle(s)
Product Line: Resurge™
Quantity: 1 kg
Form: Powder

Contents & storage

Storage conditions: 2°C to 8°C. Store in cool, dry conditions in well-sealed containers.