Soy 100 is a highly digested soy protein peptone that is high in nucleosides and free amino acid content. It has high concentrations of vitamins and carbohydrates, which is recommended for the cultivation of a wide variety of microbial organisms as well as varied mammalian cell culture applications.
Soy 100 can be blended with other animal origin-free peptones, such as wheat or yeast, when looking for replacements for animal origin peptones.
Soy 100 is also part of the Gibco Peptone Preview Pak, which features our new wheat, cotton, soy, and yeast peptones. These peptones provide unique nutritional profiles compared to other cell culture supplements. The Preview Pak includes five 100 gram samples*.
*The Gibco Peptone Preview Pak is non-GMP. Larger GMP packaging sizes are available.
Format: | Powder |
---|---|
Media Type: | Dehydrated Culture Media |
Product Line: | Gibco™ |
Quantity: | 10 kg |
Remarks: | There are no animal sourced ingredients or animal derived reagents used in the manufacture of this product. |
Solubility: | (2% and 10% solutions): Soluble in distilled or deionized water |
Shelf Life: | 2 Years |
Source: | Animal origin-free |
pH Range: | 6.2 to 7.6 |
For Use With (Application): | Cell Culture and Microbial Fermentation, Tissue Culture Application |
Packaging: | Bottle, Pail |
Regulatory Status: | For Research Use or Further Manufacturing. Not for diagnostic use or direct administration in to humans or animals |
Storage Requirements: | Store in cool, dry conditions in well-sealed container, 2°C to 25°C |