UltiMate™ AFC-3000 Adapter Blocks
Actual product may vary
UltiMate™ AFC-3000 Adapter Blocks
UltiMate™ AFC-3000 Adapter Blocks
Thermo Scientific™
UltiMate™ AFC-3000 Adapter Blocks
Add versatility to your fraction collector. These adapter blocks allow the use of well plates or sample trays in your AFC-3000 Fraction Collector.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberFor Use With (Equipment)
6702.0200Dionex AFC-3000 Automated Fraction Collector
6702.0100Ultimate AFC-3000 Fraction Collector
Catalog number 6702.0200
Price (USD)
-
For Use With (Equipment):
Dionex AFC-3000 Automated Fraction Collector
Add versatility to your fraction collector. Thermo Scientific™ Dionex™ UltiMate™ AFC-3000 Adapter Blocks allow the use of well plates or sample trays in your AFC-3000 Fraction Collector. The UltiMate AFC-3000 Automated Fraction Collector is a rugged and flexible fraction collector, ideal for preparative reversed-phase or normal phase HPLC as well as purification of proteins, peptides, and nucleotides.
Specifications
For Use With (Equipment)Dionex AFC-3000 Automated Fraction Collector
TypeAdapter Block
Unit SizeEach