Vanquish™ Fraction Collector Capillaries and Loops
Thermo Scientific™

Vanquish™ Fraction Collector Capillaries and Loops

Achieve pinpoint fractionation with precision using a Vanquish HPLC or UHPLC system with an integrated fraction collector. Choose capillaries and loops.
Have Questions?
變更視圖buttonViewtableView
產品號碼TypeFlow Rate Range
6706.1020Needle Capillary5 to 10 mL/min.
6706.1110Delay Capillary0.5 to 1 mL/min.
6706.1080Flush Buffer Loop5 to 10 mL/min.
6706.1100Delay Capillary50 μL to 0.5 mL/min.
6706.1120Delay Capillary1 to 2 mL/min.
6706.1130Delay Capillary2 to 10 mL/min.
產品號碼 6706.1020
價格 (TWD)
-
Type:
Needle Capillary
Flow Rate Range:
5 to 10 mL/min.

Modify your Thermo Scientific™ Vanquish™ Fraction Collector for dedicated fractionation application needs. Choose between different sized delay capillaries when using peak-based fractionation. For elevated flow rates select specific needle capillaries and flush buffer loops. Thermo Scientific™ Viper™ Fingertight Fittings are included with our capillaries and loops to make them easy to install with leak-free connections.

We offer different configurations to modify the default setup depending on flow rate and fractionation trigger for ultimate performance.

  • For peak-based fractionation select between different delay capillaries
  • For both peak-based and time-based fractionation above a flow rate of 5 mL/min, select the  appropriate needle capillary and flush buffer loop
規格
DescriptionNeedle capillary, 0.25 x 415 mm
Flow Rate Range5 to 10 mL/min.
MaterialPEEK-shielded MP35N
TypeNeedle Capillary
For Use With (Equipment)Vanquish Fraction Collector
Unit SizeEach