Vanquish™ Fraction Collector Capillaries and Loops
Thermo Scientific™
Vanquish™ Fraction Collector Capillaries and Loops
Achieve pinpoint fractionation with precision using a Vanquish HPLC or UHPLC system with an integrated fraction collector. Choose capillaries and loops.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberTypeFlow Rate Range
6706.1110Delay Capillary0.5 to 1 mL/min.
6706.1020Needle Capillary5 to 10 mL/min.
6706.1080Flush Buffer Loop5 to 10 mL/min.
6706.1100Delay Capillary50 μL to 0.5 mL/min.
6706.1120Delay Capillary1 to 2 mL/min.
6706.1130Delay Capillary2 to 10 mL/min.
Catalog number 6706.1110
Price (EUR)
-
Type:
Delay Capillary
Flow Rate Range:
0.5 to 1 mL/min.
Modify your Thermo Scientific™ Vanquish™ Fraction Collector for dedicated fractionation application needs. Choose between different sized delay capillaries when using peak-based fractionation. For elevated flow rates select specific needle capillaries and flush buffer loops. Thermo Scientific™ Viper™ Fingertight Fittings are included with our capillaries and loops to make them easy to install with leak-free connections.
We offer different configurations to modify the default setup depending on flow rate and fractionation trigger for ultimate performance.
  • For peak-based fractionation select between different delay capillaries
  • For both peak-based and time-based fractionation above a flow rate of 5 mL/min, select the  appropriate needle capillary and flush buffer loop
Specifications
DescriptionDelay capillary for peak-based fractionation, 0.25 x 1500 mm
Flow Rate Range0.5 to 1 mL/min.
MaterialMP35N
TypeDelay Capillary
For Use With (Equipment)Vanquish Fraction Collector
Unit SizeEach