Thermo Scientific™
nanoViper™ Kits for Preconcentration
 Related applications:
Industrial Chromatography
Maximize performance and simplify setup of your nano liquid chromatography (LC) instrument with Thermo Scientific™ nanoViper™ application kits for preconcentration. Designed specifically for use with the Thermo Scientific™ Dionex™ UltiMate™ 3000 RSLCnano system, these kits contain nanoViper fittings preassembled on connection tubing and columns to enable easy, tool-free instrument setup and configuration. The fingertight fittings ensure virtually zero-dead-volume connections and provide UHPLC compatibility up to 1000bar (14,500 psi).
Error loading your content!
  Catalog number Price ({{currency}}) Availability Qty
{{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}} Add To Cart
{{addItemToList.successMessage}} {{addItemToList.listName}}
Back to top
Description
The nanoViper Fingertight Fittings seal at the tip of the tubing to eliminate dead volumes and make it easier to plumb your HPLC or UHPLC system.
Documents
Manuals & protocols