Thermo Scientific™
nanoViper™ Tandem nanoLC Kit for UltiMate™ 3000 RSLCnano system
 Related applications:
Industrial Chromatography
Avoid data-destroying dead volumes of your tandem nanoLC system by using the Thermo Scientific™ Dionex™ nanoViper™ Tandem nanoLC Kit. Viper Fingertight Fittings seal at the tip of the tubing to eliminate dead volumes and ensure easy, perfect connections in your HPLC or UHPLC system. A single kit contains all the connectors and capillaries you need and ensures consistently perfect connections.
Error loading your content!
  Catalog number Price ({{currency}}) Availability Qty
{{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}} Add To Cart
{{addItemToList.successMessage}} {{addItemToList.listName}}
Back to top
Description
nanoViper fittings are an integrated part of the nano and capillary workflows and eliminate the assembly of PEEK sleeve connections. This fingertight system comes preassembled on connection tubings and columns to ensure system setup is never difficult.
Provides Virtually Zero-Dead-Volume Connections
  • Available in many internal diameters and lengths
  • Available individually or as complete application kits
Easy, Worry-Free Connections
  • Supports backpressures up to 100MPa (1,000bar/12,600psi)
  • Suitable for temperatures up to 80°C
  • Works with virtually any valve, and any column, from any manufacturer
  • Easy to use with fingertight design
  • Paves the way for the easy, tool-free nano LC setup of any application
Documents
Manuals & protocols