Thermo Scientific™
nanoViper™ Automated Offline Column Peptide Kit
Related applications:
Industrial & Applied Science
Maximize performance and simplify setup of your nano liquid chromatography (LC) instrument with Thermo Scientific™ Dionex™ nanoViper™ Automated Offline Column Peptide Kit. Designed specifically for use with the Thermo Scientific™ Dionex™ UltiMate™ 3000 RSLCnano system, these kits contain nanoViper fittings preassembled on connection tubing and columns to enable easy, tool-free instrument setup and configuration. The fingertight fittings ensure virtually zero-dead-volume connections and provide UHPLC compatibility up to 1000 bar (14,500 psi).
Back to top
The nanoViper Fingertight Fittings seal at the tip of the tubing to eliminate dead volumes and make it easier to plumb your HPLC or UHPLC system. The nanoViper fittings are an integrated part of the nano and capillary workflows and eliminate the assembly of PEEK sleeve connections. This fingertight system comes preassembled on connection tubings and columns to ensure system setup is never difficult.
Provides Virtually Zero-Dead-Volume Connections
  • Available in many internal diameters and lengths
  • Available individually or as complete application kits
Easy, Worry-Free Connections
  • Supports backpressures up to 100MPa (1,000bar/12,600psi)
  • Suitable for temperatures up to 80°C
  • Works with virtually any valve, and any column, from any manufacturer
  • Easy to use with fingertight design
  • Paves the way for the easy, tool-free nano LC setup of any application