Thermo Scientific™
UltiMate™ TCC-3000RS/SD SST Valve Pods
Related applications:
Industrial Chromatography
Choose Thermo Scientific™ Dionex™ UltiMate™ TCC-3000RS/SD SST Valve Pods for Two-Position HP Valves for use with the TCC-3000RS/SD Thermostatted Column Compartment.
Back to top
  • Stainless-steel
  • Pressure <413 bar (5000psi)