Vanquish™ Passive Pre-Heaters TQ
Thermo Scientific™
Vanquish™ Passive Pre-Heaters TQ
Contribute to improved resolution and retention time stability in the Thermo Scientific™ Vanquish™ UHPLC system with active or passive eluent pre-heaters using Thermo Scientific™ Viper™ TQ UHPLC Fitting technology.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberLength (Metric)Sample CapacityMaterial
6732.0750580 mm5 μLStainless Steel
6732.0740580 mm3 μLStainless Steel
6732.0760580 mm1 μLMP35N
6732.0780730 mm1 μLMP35N
Catalog number 6732.0750
Price (MXN)
-
Length (Metric):
580 mm
Sample Capacity:
5 μL
Material:
Stainless Steel
Ensure precise chromatographic performance in the Thermo Scientific™ Vanquish™ UHPLC system with the Thermo Scientific™ Active Pre-heater TQ or Passive Pre-heater TQ. The pre-heaters come with torque-controlled Viper TQ UHPLC Fittings for confident, leak-free connections with audible and tactile feedback to be installed in Thermo Scientific™ Vanquish™ Column Compartment H or C.
The Active Pre-heater matches the temperature of solvent entering the column with the temperature of the column thereby avoiding performance loss from temperature effects. The Active Pre-heater TQ has separate power, heating coil, and temperature controls and comes with replaceable outlet capillaries to extend the lifespan of the part.
Optimizing chromatographic performance with eluent pre-heating
  • Easy to install with the Thermo Scientific™ Viper™ TQ UHPLC fitting technology
  • Reduced dispersion and optimal heat transfer with dedicated formats tailored for the Vanquish HPLC and UHPLC flow paths
  • Wide application flexibility from HPLC to UHPLC using different materials and geometric dimensions
Active pre-heater TQ
The active pre-heater has separate power and a heating coil. The independent temperature control allows the fine-tuning of the eluent temperature for reliable chromatographic methods, method development or method transfer. The replaceable outlet capillaries reduce cost-of-ownership and enhance sustainability with the Active pre-heater TQ beside offering versatile column compatibility.
Passive pre-heater
The passive pre-heater allows for heating and cooling of the eluent to match the column temperature.
Specifications
Length (Metric)580 mm
For Use With (Equipment)Vanquish Column Compartments
MaterialStainless Steel
Max. Pressure1517 bar
Sample Capacity5 μL
TypePre-heater
Unit SizeEach