Thermo Scientific™
Shandon™ TBD-1™ Decalcifier
Related applications:
Anatomical Pathology
Process bone marrow biopsies in as little as 15 to 30 minutes with the Thermo Scientific™ Shandon™ TBD-1™ Decalcifier, a rapid hydrochloric acid decalcifier.
Back to top
  • Process thin pieces of dense bone in 3 to 12 hours
  • Permits utilization of routine end point tests, such as calcium oxalate
  • Protects cells from actions of acids with added ingredients