Thermo Scientific™
Shandon™ Zinc Formal-Fixx™
Related applications:
Anatomical Pathology
Acquire amazing results with Thermo Scientific™ Shandon™ Zinc Formal-Fixx™, which is excellent for immunohistochemistry.
Back to top
  • Enhances H&E's and special stains
  • Minimizes the need to enzyme digest
  • Reduces protein cross-linking
  • Produces well-defined nuclear detail