Thermo Scientific™
Shandon™ Glyo-Fixx™
Related applications:
Anatomical Pathology
Obtain superior results to neutral buffered formalin with Thermo Scientific™ Shandon™ Glyo-Fixx™, a noncarcinogenic, nonsensitizing, and environmentally friendly substitute.
Back to top
  • Produces distinct and crisp cellular morphology
  • Fixes most antigens without impairing immunoreactivity
  • Allows special stain dyes, including silver procedures, to be sharp without background staining
  • Presents fine detail in specimen structure