Thermo Scientific™
Shandon™ EA-50 Stain
Related applications:
Anatomical Pathology
Clearly differentiate cellular components with pink, green and blue-green hues utilizing Thermo Scientific™ Shandon™ EA-50 Stain.
Back to top
  • Polychromatic cytoplasmic stain
  • Biological stain commission-certified dye