Thermo Scientific™
Shandon™ Acid-Resistant Biopsy Bags
Related applications:
Anatomical Pathology
Reduce risk of small specimen loss with Thermo Scientific™ Shandon™ Acid-Resistant Biopsy Bags. These bags feature all the benefits of nylon biopsy bags, plus offer acid-resistance for decalcification.
Back to top
  • Allow unrestricted fluid movement around tissue
  • Reduce reagent carryover and pressure artifact often found with sponges
  • Remove specimen rapidly with convenient peel-open bag
Dimensions:
  • Small Biopsy Bags measure 1.2 × 1.7 in. (31 × 43mm)
  • Medium Biopsy Bags measure 1.8 × 2.6 in. (45 × 65mm)
  • Large Biopsy Bags measure 3 × 3.5 in. (75 × 85mm)