SST Buffer Loops for Split Loop Autosamplers
Product Image
Thermo Scientific™
SST Buffer Loops for Split Loop Autosamplers
Order the stainless steel Thermo Scientific™ Dionex™ Buffer Loops (100 and greater than 250μL) you need the UltiMate™ split loop autosamplers.
Have Questions?
Alterar visualizaçãobuttonViewtableView
Número do catálogoSample Capacity
6820.2468>250 μL
6820.2466100 μL
Número do catálogo 6820.2468
Preço (BRL)
-
Sample Capacity:
>250 μL
Order the stainless steel Thermo Scientific™ Dionex™ Buffer Loops (100 and greater than 250μL) you need the Thermo Scientific™ Dionex™ UltiMate™ split loop autosamplers. The sample loops incorporate Thermo Scientific™ Dionex™ Viper™ Fingertight Fittings which eliminate dead volumes and ensure easy, perfect connections in your HPLC or UHPLC system every time.
Especificações
For Use With (Equipment)Dionex UltiMate 3000 Split Loop Autosampler WPS-3000(T)SL Semiprep
MaterialStainless Steel
Sample Capacity>250 μL
TypeBuffer Loop
Unit SizeEach