Sample Racks for UltiMate™ 3000 Autosamplers, Column Compartments, and Fraction Collectors
Imagem ilustrativa. O produto pode variar
Thermo Scientific™
Sample Racks for UltiMate™ 3000 Autosamplers, Column Compartments, and Fraction Collectors
Sample Racks are used for UltiMate™ 3000 Autosamplers, Column Compartments, and Fraction Collectors.
Have Questions?
Alterar visualizaçãobuttonViewtableView
Número do catálogoFor Use With (Equipment)
6820.408610 mL Vials
6820.40701.8 mL Vials
6820.40844 mL Vials
6820.40871.1 mL Vials
6820.40901.2 mL Vials
6820.40910.3 mL Vials
6820.40941.5 mL Eppendorf Tubes
6820.40960.5 mL Eppendorf Tubes
6820.40970.3 mL Micro-Dialyse Vials
Número do catálogo 6820.4086
Preço (BRL)
-
For Use With (Equipment):
10 mL Vials
Support exceptional sample format versatility with Thermo Scientific™ Dionex™ Sample Racks used for UltiMate™ 3000 Autosamplers, Column Compartments, and Fraction Collectors, for the WPS-3000 series UHPLC and LC samplers.
Especificações
Descriptionfor 10 vials, 10 mL, cylindrical
Sample Capacity10 mL
For Use With (Equipment)10 mL Vials
Product LineDionex
TypeAutosampler Rack
Unit SizeEach