Product Image
Thermo Scientific™
Well Plate Support Racks for UltiMate™ Autosamplers and Column Compartments
Well Plate Support Racks for UltiMate™ Autosamplers and Column Compartments are for use with ACC- and WPS-3000 autosamplers.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDescription
6820.408920-32 mm height plate, 96 or 384 wells
6820.407934-46 mm height plates
6820.408330-36 mm height plates
6820.408810-12 mm height plates
Catalog number 6820.4089
Price (HKD)
-
Description:
20-32 mm height plate, 96 or 384 wells
Use Thermo Scientific™ Dionex™ Well Plate Support Racks for UltiMate™ Autosamplers and Column Compartments with ACC- and WPS-3000 autosamplers.
Specifications
Description20-32 mm height plate, 96 or 384 wells
For Use With (Equipment)Deep-Well Plates for All WPS-3000 LC Autosamplers (Not ACC)
Sample Capacity96 or 384 Wells
TypeLow Profile Support Rack
Unit SizeEach