Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Type | Buffer Tubing |
Sample Capacity | 50 μL |
Max. Pressure | 345 bar |
Biocompatible Version | Yes |
For Use With (Equipment) | Ultimate 3000 Autosamplers WPS-3000 TBPL Nano/Cap and WPS-3000 TBFC Analytical Optimized for Nano/Cap |
Unit Size | Each |
Type | Buffer Tubing |
Sample Capacity | 1000 μL |
Biocompatible Version | Yes |
For Use With (Equipment) | WPS-3000 TBPL Analytical and WPS-3000 TBFC Analytical |
Unit Size | - |
Type | Buffer Tubing |
Sample Capacity | 2000 μL |
Biocompatible Version | Yes |
For Use With (Equipment) | Ultimate 3000 Autosampler WPS-3000 TBPL Nano/Cap |
Unit Size | Each |
Type | Buffer Tubing |
Sample Capacity | 500 μL |
Biocompatible Version | Yes |
For Use With (Equipment) | Ultimate 3000 Autosamplers WPS-3000 TBPL Nano/Cap, WPS-3000 TBPL Analytical and WPS-3000 TBFC Analytical |
Unit Size | Each |
Catalog Number | Specifications | Unit Size | For Use With (Equipment) | Type | Sample Capacity | Price (USD) | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|
6821.0019 | Each | Ultimate 3000 Autosamplers WPS-3000 TBPL Nano/Cap and WPS-3000 TBFC Analytical Optimized for Nano/Cap | Buffer Tubing | 50 μL | Request A Quote | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
6821.0022 | - | WPS-3000 TBPL Analytical and WPS-3000 TBFC Analytical | Buffer Tubing | 1000 μL | Request A Quote | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
6821.0021 | Each | Ultimate 3000 Autosampler WPS-3000 TBPL Nano/Cap | Buffer Tubing | 2000 μL | Request A Quote | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
6821.0020 | Each | Ultimate 3000 Autosamplers WPS-3000 TBPL Nano/Cap, WPS-3000 TBPL Analytical and WPS-3000 TBFC Analytical | Buffer Tubing | 500 μL | Request A Quote | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
|