Product Image
Thermo Scientific™

Buffer Tubing for UltiMate™ 3000 Autosamplers, Biocompatible

Select this biocompatible version of buffer tubing in volumes from 50 to 2000μL for your biocompatible Thermo Scientific™ UltiMate™ 3000 Autosampler.
Have Questions?
變更視圖buttonViewtableView
產品號碼Sample Capacity
6821.0020500 μL
6821.00221000 μL
6821.00212000 μL
6821.001950 μL
產品號碼 6821.0020
價格 (HKD)
-
Sample Capacity:
500 μL

Choose Thermo Scientific™ Dionex™ Biocompatible Buffer Tubing for Thermo Scientific™ UltiMate™ 3000 Autosamplers, available in volumes from 50 to 2000μL. Select the biocompatible buffer tubing volume of choice your biocompatible LC system.

規格
Biocompatible VersionYes
For Use With (Equipment)Ultimate 3000 Autosamplers WPS-3000 TBPL Nano/Cap, WPS-3000 TBPL Analytical and WPS-3000 TBFC Analytical
Sample Capacity500 μL
TypeBuffer Tubing
Unit SizeEach