Product Image
Thermo Scientific™

Buffer Tubing for UltiMate™ 3000 Autosamplers, Biocompatible

Select this biocompatible version of buffer tubing in volumes from 50 to 2000μL for your biocompatible Thermo Scientific™ UltiMate™ 3000 Autosampler.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberSample Capacity
6821.00221000 μL
6821.00212000 μL
6821.001950 μL
6821.0020500 μL
Catalog number 6821.0022
Price (EUR)
-
Sample Capacity:
1000 μL

Choose Thermo Scientific™ Dionex™ Biocompatible Buffer Tubing for Thermo Scientific™ UltiMate™ 3000 Autosamplers, available in volumes from 50 to 2000μL. Select the biocompatible buffer tubing volume of choice your biocompatible LC system.

Specifications
QuantityEach
Biocompatible VersionYes
For Use With (Equipment)WPS-3000 TBPL Analytical and WPS-3000 TBFC Analytical
Sample Capacity1000 μL
TypeBuffer Tubing
Unit SizeEach