Syringes for UltiMate™ 3000 Autosamplers and Column Compartments
Actual product may vary
Thermo Scientific™
Syringes for UltiMate™ 3000 Autosamplers and Column Compartments
Available in 25, 100, 250, 500, 1000, and 2500μL capacities, the Thermo Scientific™ Dionex™ Syringes for UltiMate™ 3000 Autosamplers and Column Compartments are for use with WPS-3000 autosamplers and ACC-3000 column compartments.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberFor Use With (Equipment)Sample Capacity
6822.0005All WPS-3000 Autosamplers and ACC-3000 Column Compartments1000 μL
6822.0001All WPS-3000 Series Autosamplers25 μL
6822.0002All WPS-3000 Series Autosamplers100 μL
6822.0004All WPS-3000 Autosamplers, Except WPS-3000 TXRS500 μL
6822.0003All WPS-3000 Autosamplers and ACC-3000 Column Compartments250 μL
6822.0006WPS-3000(T)SL Semiprep Autosampler2500 μL
Catalog number 6822.0005
Price (TWD)
-
For Use With (Equipment):
All WPS-3000 Autosamplers and ACC-3000 Column Compartments
Sample Capacity:
1000 μL
Available in 25, 100, 250, 500, 1000, and 2500μL capacities, the Thermo Scientific™ Dionex™ Syringes for UltiMate™ 3000 Autosamplers and Column Compartments are for use with WPS-3000 autosamplers and ACC-3000 column compartments.
Specifications
Description1000 μL syringe for all WPS-3000 and ACC-3000 variants
Biocompatible VersionNo
For Use With (Equipment)All WPS-3000 Autosamplers and ACC-3000 Column Compartments
Sample Capacity1000 μL
TypeAutosampler Syringe
Unit SizeEach