Thermo Scientific™
UltiMate™ 3000 Split Loop Autosamplers Injection Volume Kits
Related applications:
Industrial Chromatography
Change or order new injection loops when your applications demand. Thermo Scientific™ Dionex™ UltiMate™ 3000 Split Loop Autosampler Injection Volume Kits. They are available in 250, 500 and 1000µL and include Thermo Scientific™ Dionex™ Viper™ Fingertight Fittings to ensure dead-volume-free connections.
Back to top
Description
Change or order new injection loops when your applications demand. Thermo Scientific™ Dionex™ UltiMate™ 3000 Split Loop Autosampler Injection Volume Kits. They are available in 250, 500 and 1000µL and include Thermo Scientific™ Dionex™ Viper™ Fingertight Fittings to ensure dead-volume-free connections.