PEEK Biocompatible Sample Loops for Pulled Loop Autosamplers
Actual product may vary
Thermo Scientific™
PEEK Biocompatible Sample Loops for Pulled Loop Autosamplers
Order the Thermo Scientific™ Biocompatible Sample Loops (5-250μL) you need for Thermo Scientific™ Dionex™ UltiMate™ WPS-3000TBPL pulled loop autosamplers.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberFor Use With (Equipment)Sample Capacity
6823.0019Dionex UltiMate 3000 Autosampler WPS-3000 TBPL Analytical, Dionex UltiMate 3000 Autosampler WPS-3000 TBFC Analytical50 μL
6821.0032Dionex UltiMate 3000 Autosampler WPS-3000 TBPL Analytical125 μL
6823.0016Dionex UltiMate 3000 Autosampler WPS-3000 TBPL Analytical5 μL
6823.0017Dionex UltiMate 3000 Autosampler WPS-3000 TBPL Analytical10 μL
6823.0018Dionex UltiMate 3000 Autosampler WPS-3000 TBPL Analytical20 μL
6823.0020Dionex UltiMate 3000 Autosampler WPS-3000 TBPL Analytical, Dionex UltiMate 3000 Autosampler WPS-3000 TBFC Analytical250 μL
Catalog number 6823.0019
Price (MXN)
-
For Use With (Equipment):
Dionex UltiMate 3000 Autosampler WPS-3000 TBPL Analytical, Dionex UltiMate 3000 Autosampler WPS-3000 TBFC Analytical
Sample Capacity:
50 μL
Order the Thermo Scientific™ Biocompatible Sample Loops (5-250μL) you need for Thermo Scientific™ Dionex™ UltiMate™ WPS-3000TBPL pulled loop autosamplers.
Specifications
Biocompatible VersionYes
For Use With (Equipment)Dionex UltiMate 3000 Autosampler WPS-3000 TBPL Analytical, Dionex UltiMate 3000 Autosampler WPS-3000 TBFC Analytical
MaterialPEEK
Max. Pressure345 bar
Sample Capacity50 μL
TypeSample Loop
Unit SizeEach