Product Image
Thermo Scientific™
SST Sample Loops for UltiMate™ WPS-3000TFC Pulled-Loop Autosampler
Order the stainess steel sample loop (50 and 250μL) you need for use in the Thermo Scientific™ Dionex™ UltiMate™ WPS-3000TFC pulled-loop autosampler.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberSample Capacity
6824.001950 μL
6824.0020250 μL
Catalog number 6824.0019
Price (MXN)
-
Sample Capacity:
50 μL
Order the Thermo Scientific™ SST Sample Loops you need for use in the Thermo Scientific™ Dionex™ UltiMate™ WPS-3000TFC pulled-loop autosampler. These stainless steel (SST) sample loops are available in 25 and 250μL sizes.
Specifications
For Use With (Equipment)Dionex UltiMate 3000 Well Plate Autosampler WPS-3000 TFC Analytical
MaterialStainless Steel
Sample Capacity50 μL
TypeSample Loop
Unit SizeEach